Assay Details
| Gene Symbol: Cysltr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281859.1 | NP_001268788.1 | 3 - 4 | 610 | 82 | |
| NM_021476.5 | NP_067451.2 | 3 - 4 | 629 | 82 | ||
| GenBank mRNA | AB044087.1 | - | 3 - 4 | 540 | 82 | |
| AF205830.1 | - | 3 - 4 | 418 | 82 | ||
| AF263370.1 | - | 3 - 4 | 530 | 82 | ||
| AK033476.1 | - | 3 - 4 | 413 | 82 | ||
| BC027102.1 | - | 3 - 4 | 433 | 5012266 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB044087.1, AF205830.1, AF263370.1, AK033476.1, BC027102.1, NM_001281859.1, NM_021476.5 transcripts.
Set Membership: |
|

Cysltr1
Genomic Map