Assay Details
| Gene Symbol: Taf6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146092.2 | NP_666204.2 | 7 - 8 | 709 | 77 | |
| GenBank mRNA | AK144881.1 | - | 7 - 8 | 676 | 77 | |
| BC020299.1 | - | 7 - 8 | 682 | 77 | ||
| BC028647.1 | - | 7 - 8 | 656 | 1244243 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf6l
Genomic Map