Assay Details
| Gene Symbol: Hnrnpul2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081196.1 | NP_001074665.1 | 12 - 13 | 2928 | 87 | |
| GenBank mRNA | DQ000354.1 | - | 12 - 13 | 2047 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpul2
Genomic Map