Assay Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276704.1 | NP_001263633.1 | 10 - 11 | 2933 | 74 | |
| NM_016813.2 | NP_058093.2 | 11 - 12 | 1182 | 74 | ||
| GenBank mRNA | AF093140.1 | - | 11 - 12 | 1158 | 74 | |
| AK052937.1 | - | 1 - 2 | 349 | 74 | ||
| AK077644.1 | - | 11 - 12 | 1139 | 74 | ||
| AK079583.1 | - | 11 - 12 | 1171 | 74 | ||
| AK086527.1 | - | 10 - 11 | 2944 | 74 | ||
| AK088634.1 | - | 10 - 11 | 2904 | 74 | ||
| AK133883.1 | - | 11 - 12 | 1171 | 74 | ||
| AK143640.1 | - | 2934 | 74 | |||
| AK167518.1 | - | 11 - 12 | 1170 | 74 | ||
| AK191466.1 | - | 2 - 3 | 138 | 74 | ||
| BC005594.1 | - | 11 - 12 | 1182 | 3710546 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf1
Genomic Map