Assay Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276704.1 | NP_001263633.1 | 4 - 5 | 564 | 101 | |
| NM_016813.2 | NP_058093.2 | 4 - 5 | 578 | 101 | ||
| GenBank mRNA | AF093140.1 | - | 4 - 5 | 554 | 101 | |
| AK077644.1 | - | 4 - 5 | 535 | 101 | ||
| AK086527.1 | - | 4 - 5 | 575 | 101 | ||
| AK088634.1 | - | 4 - 5 | 535 | 101 | ||
| AK133883.1 | - | 4 - 5 | 567 | 101 | ||
| AK143640.1 | - | 565 | 101 | |||
| AK167518.1 | - | 4 - 5 | 566 | 101 | ||
| BC005594.1 | - | 4 - 5 | 578 | 3710546 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf1
Genomic Map