Assay Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276704.1 | NP_001263633.1 | 7 - 8 | 818 | 83 | |
| NM_016813.2 | NP_058093.2 | 7 - 8 | 832 | 83 | ||
| GenBank mRNA | AF093140.1 | - | 7 - 8 | 808 | 83 | |
| AK077644.1 | - | 7 - 8 | 789 | 83 | ||
| AK086527.1 | - | 7 - 8 | 829 | 83 | ||
| AK088634.1 | - | 7 - 8 | 789 | 83 | ||
| AK133883.1 | - | 7 - 8 | 821 | 83 | ||
| AK143640.1 | - | 819 | 83 | |||
| AK167518.1 | - | 7 - 8 | 820 | 83 | ||
| BC005594.1 | - | 7 - 8 | 832 | 3710546 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf1
Genomic Map