Assay Details
| Gene Symbol: Mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199274.1 | NP_001186203.1 | 1 - 2 | 403 | 85 | |
| GenBank mRNA | AK075947.1 | - | 1 - 2 | 142 | 85 | |
| AK162087.1 | - | 1 - 2 | 142 | 85 | ||
| CA559663.1 | - | 1 - 2 | 403 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK075947.1, AK162087.1, NM_001199274.1 transcripts.
Set Membership: |
|

Mat2b
Genomic Map