Assay Details
| Gene Symbol: Eml3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144872.1 | NP_659121.1 | 8 - 9 | 1240 | 82 | |
| GenBank mRNA | BC022146.1 | - | 8 - 9 | 1240 | 5101626 | 82 |
| BC023878.1 | - | 8 - 9 | 1263 | 5354867 | 82 | |
| BC034112.1 | - | 8 - 9 | 1038 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eml3
Genomic Map