Assay Details
| Gene Symbol: Snhg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK051045.1 | - | 2 - 3 | 1827 | 150 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 22 |
Snord22 |
Rnu22, U22 |
Sense | Chr. 19 - 8725866 - 8725991 on NCBI Build GRCm38 | |
| Downstream | WD repeat domain 74 | Wdr74 |
5730436H21Rik, AA407588 |
9.7kb | Sense | Chr. 19 - 8735799 - 8740624 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Snhg1
Genomic Map