Assay Details
| Gene Symbol: Chka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271496.1 | NP_001258425.1 | 9 - 10 | 1411 | 107 | |
| NM_013490.4 | NP_038518.2 | 8 - 9 | 1357 | 107 | ||
| NR_073190.1 | 10 - 11 | 1471 | 107 | |||
| GenBank mRNA | AB011002.1 | - | 8 - 9 | 1216 | 107 | |
| AK166465.1 | - | 8 - 9 | 1298 | 107 | ||
| AK184298.1 | - | 2 - 3 | 151 | 107 | ||
| BC004719.1 | - | 6 - 7 | 547 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chka
Genomic Map