Assay Details
| Gene Symbol: Tcirg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136091.2 | NP_001129563.1 | 3 - 4 | 316 | 54 | |
| NM_001167784.1 | NP_001161256.1 | 3 - 4 | 661 | 54 | ||
| NM_016921.3 | NP_058617.3 | 3 - 4 | 357 | 54 | ||
| GenBank mRNA | AB022322.1 | - | 3 - 4 | 223 | 54 | |
| AK155140.1 | - | 3 - 4 | 662 | 54 | ||
| BC006761.1 | - | 3 - 4 | 357 | 3257437 | 54 | |
| BC085234.1 | - | 3 - 4 | 217 | 5322429 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcirg1
Genomic Map