Assay Details
| Gene Symbol: Itgb1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008403.4 | NP_032429.1 | 5 - 6 | 577 | 91 | |
| GenBank mRNA | AJ001373.1 | - | 5 - 6 | 559 | 91 | |
| AK002786.1 | - | 5 - 6 | 534 | 91 | ||
| AK004172.1 | - | 5 - 6 | 533 | 91 | ||
| AK007635.1 | - | 5 - 6 | 535 | 91 | ||
| AK075577.1 | - | 5 - 6 | 578 | 91 | ||
| AK089937.1 | - | 5 - 6 | 691 | 91 | ||
| AK147929.1 | - | 5 - 6 | 526 | 91 | ||
| BC021151.1 | - | 5 - 6 | 716 | 91 | ||
| BC028772.1 | - | 4 - 5 | 413 | 1230640 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb1bp1
Genomic Map