Assay Details
| Gene Symbol: Dach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038610.2 | NP_001033699.1 | 5 - 6 | 1873 | 100 | |
| NM_007826.3 | NP_031852.1 | 6 - 7 | 2029 | 100 | ||
| GenBank mRNA | AF090436.1 | - | 6 - 7 | 1573 | 100 | |
| AF090437.1 | - | 5 - 6 | 1417 | 100 | ||
| AF102547.1 | - | 6 - 7 | 1573 | 100 | ||
| AF129510.1 | - | 5 - 6 | 1635 | 100 | ||
| AJ005669.1 | - | 6 - 7 | 1091 | 100 | ||
| AK047409.1 | - | 5 - 6 | 739 | 100 | ||
| AK053594.1 | - | 5 - 6 | 1082 | 100 | ||
| AK139060.1 | - | 5 - 6 | 1874 | 100 | ||
| BC052005.1 | - | 4 - 5 | 1117 | 100 | ||
| BC059934.1 | - | 5 - 6 | 1651 | 100 | ||
| BC078644.1 | - | 5 - 6 | 1407 | 100 | ||
| BC141130.1 | - | 5 - 6 | 1479 | 9055742 | 100 | |
| BC145440.1 | - | 5 - 6 | 1479 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dach1
Genomic Map