Assay Details
| Gene Symbol: Rce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023131.1 | NP_075620.1 | 3 - 4 | 378 | 63 | |
| GenBank mRNA | AJ251645.1 | - | 3 - 4 | 378 | 63 | |
| AK012946.1 | - | 3 - 4 | 334 | 63 | ||
| BC004060.1 | - | 2 - 3 | 227 | 3590170 | 63 | |
| BC048686.1 | - | 3 - 4 | 402 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rce1
Genomic Map