Assay Details
| Gene Symbol: Pola2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164057.1 | NP_001157529.1 | 4 - 5 | 660 | 95 | |
| NM_008893.3 | NP_032919.2 | 4 - 5 | 660 | 95 | ||
| GenBank mRNA | AK087989.1 | - | 4 - 5 | 656 | 95 | |
| AK133832.1 | - | 4 - 5 | 646 | 95 | ||
| AK150099.1 | - | 4 - 5 | 602 | 95 | ||
| AK152974.1 | - | 4 - 5 | 624 | 95 | ||
| AK153081.1 | - | 4 - 5 | 627 | 95 | ||
| AK163733.1 | - | 4 - 5 | 598 | 95 | ||
| BC006945.1 | - | 4 - 5 | 606 | 3602559 | 95 | |
| BC021424.1 | - | 4 - 5 | 635 | 5011204 | 95 | |
| BC023688.1 | - | 4 - 5 | 674 | 5343583 | 95 | |
| BC064795.1 | - | 4 - 5 | 576 | 30111287 | 95 | |
| D13546.1 | - | 4 - 5 | 593 | 95 | ||
More Information
Set Membership: |
|

Pola2
Genomic Map