Assay Details
| Gene Symbol: Capn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110504.1 | NP_001103974.1 | 15 - 16 | 1759 | 79 | |
| NM_007600.3 | NP_031626.1 | 15 - 16 | 1811 | 79 | ||
| GenBank mRNA | AF021847.1 | - | 15 - 16 | 1690 | 79 | |
| AF084459.1 | - | 14 - 15 | 1677 | 79 | ||
| AK088547.1 | - | 15 - 16 | 1731 | 79 | ||
| AK149090.1 | - | 11 - 12 | 1159 | 79 | ||
| AK149706.1 | - | 15 - 16 | 1752 | 79 | ||
| AK150084.1 | - | 15 - 16 | 1750 | 79 | ||
| AK160940.1 | - | 15 - 16 | 1751 | 79 | ||
| AK165258.1 | - | 15 - 16 | 1771 | 79 | ||
| AK168063.1 | - | 15 - 16 | 1750 | 79 | ||
| AK171405.1 | - | 15 - 16 | 1714 | 79 | ||
| BC026138.1 | - | 15 - 16 | 1726 | 4218148 | 79 | |
| BC050276.1 | - | 15 - 16 | 1727 | 4923347 | 79 | |
More Information
Set Membership: |
|

Capn1
Genomic Map