Assay Details
| Gene Symbol: Tm7sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028454.2 | NP_082730.2 | 2 - 3 | 382 | 67 | |
| GenBank mRNA | AF480070.1 | - | 2 - 3 | 256 | 67 | |
| AK039028.1 | - | 2 - 3 | 312 | 67 | ||
| AK040016.1 | - | 2 - 3 | 388 | 67 | ||
| AK043687.1 | - | 2 - 3 | 291 | 67 | ||
| AK155475.1 | - | 2 - 3 | 339 | 67 | ||
| BC014769.1 | - | 2 - 3 | 374 | 4239159 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm7sf2
Genomic Map