Assay Details
| Gene Symbol: Tm7sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028454.2 | NP_082730.2 | 6 - 7 | 852 | 90 | |
| GenBank mRNA | AF480070.1 | - | 6 - 7 | 726 | 90 | |
| AK014166.1 | - | 6 - 7 | 833 | 90 | ||
| AK155475.1 | - | 6 - 7 | 809 | 90 | ||
| BC014769.1 | - | 6 - 7 | 844 | 4239159 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm7sf2
Genomic Map