Assay Details
| Gene Symbol: 4933400C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040770.1 | 1 - 2 | 369 | 66 | ||
| GenBank mRNA | AK077069.1 | - | 1 - 2 | 370 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
procollagen lysine, 2-oxoglutarate 5-dioxygenase 2 |
Plod2 |
D530025C14Rik, LH2, Plod-2 |
-110.3kb | Sense | Chr. 9 - 92542223 - 92608428 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700034K08 gene | 1700034K08Rik | 206.4kb | Anti-sense | Chr. 9 - 92979916 - 93005733 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0800001298.rn
Set Membership: |
|

4933400C23Rik
Genomic Map