Assay Details
| Gene Symbol: Rasgrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011242.2 | NP_035372.2 | 11 - 12 | 1439 | 62 | |
| GenBank mRNA | AK034683.1 | - | 5 - 6 | 691 | 62 | |
| AK160563.1 | - | 1 - 2 | 170 | 62 | ||
| Y12339.1 | - | 3 - 4 | 258 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rasgrp2
Genomic Map