Assay Details
| Gene Symbol: Rasgrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011242.2 | NP_035372.2 | 14 - 15 | 1733 | 62 | |
| GenBank mRNA | AK160563.1 | - | 4 - 5 | 464 | 62 | |
| Y12339.1 | - | 6 - 7 | 552 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rasgrp2
Genomic Map