Assay Details
| Gene Symbol: Rasgrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011242.2 | NP_035372.2 | 2 - 3 | 212 | 56 | |
| GenBank mRNA | BC027446.1 | - | 1 - 2 | 1751 | 56 | |
| BC051474.1 | - | 1 - 2 | 171 | 56 | ||
| CJ057054.1 | - | 2 - 3 | 213 | 56 | ||
| CO425553.1 | - | 1 - 2 | 137 | 56 | ||
More Information
Set Membership: |
|

Rasgrp2
Genomic Map