Assay Details
| Gene Symbol: Rasgrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011242.2 | NP_035372.2 | 5 - 6 | 509 | 58 | |
| GenBank mRNA | CO425553.1 | - | 4 - 5 | 434 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasgrp2
Genomic Map