Assay Details
| Gene Symbol: Rasgrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011242.2 | NP_035372.2 | 7 - 8 | 841 | 69 | |
| GenBank mRNA | AK034683.1 | - | 1 - 2 | 93 | 69 | |
| CO425553.1 | - | 6 - 7 | 765 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rasgrp2
Genomic Map