Assay Details
| Gene Symbol: Rasgrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011242.2 | NP_035372.2 | 9 - 10 | 1237 | 82 | |
| GenBank mRNA | AK034683.1 | - | 3 - 4 | 489 | 82 | |
| Y12339.1 | - | 1 - 2 | 56 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rasgrp2
Genomic Map