Assay Details
| Gene Symbol: Rin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145495.2 | NP_663470.1 | 6 - 7 | 1300 | 64 | |
| GenBank mRNA | BC011277.1 | - | 6 - 7 | 1300 | 3711198 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|

Rin1
Genomic Map