Assay Details
| Gene Symbol: C430002N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102296.1 | 3 - 4 | 384 | 105 | ||
| GenBank mRNA | AK049386.1 | - | 3 - 4 | 372 | 105 | |
| AK089981.1 | - | 374 | 105 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA E030011O05 gene | E030011O05Rik | -17.1kb | Anti-sense | Chr. 9 - 96731575 - 96752831 on NCBI Build GRCm38 | |
| Downstream | 2-phosphoxylose phosphatase 1 | Pxylp1 |
9430094M07Rik, Acpl2, BB177120, C130099A20Rik |
51.7kb | Anti-sense | Chr. 9 - 96823336 - 96892680 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

C430002N11Rik
Genomic Map