Assay Details
| Gene Symbol: Fkbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166368.1 | NP_001159840.1 | 3 - 4 | 345 | 70 | |
| NM_008020.3 | NP_032046.1 | 3 - 4 | 360 | 70 | ||
| GenBank mRNA | AK144284.1 | - | 3 - 4 | 361 | 70 | |
| AK165394.1 | - | 3 - 4 | 325 | 70 | ||
| BC031824.1 | - | 3 - 4 | 314 | 4196363 | 70 | |
| BC053692.1 | - | 3 - 4 | 350 | 6395841 | 70 | |
| CF949699.1 | - | 2 - 3 | 164 | 70 | ||
| DV655383.1 | - | 3 - 4 | 345 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp2
Genomic Map