Assay Details
| Gene Symbol: Pxylp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289645.1 | NP_001276574.1 | 1 - 2 | 120 | 87 | |
| GenBank mRNA | AK036973.1 | - | 1 - 2 | 121 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036973.1, NM_001289645.1 transcripts.
Set Membership: |
|

Pxylp1
Genomic Map