Assay Details
| Gene Symbol: Col6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026763.2 | NP_081039.2 | 17 - 18 | 4572 | 131 | |
| GenBank mRNA | AB370265.1 | - | 17 - 18 | 4664 | 131 | |
| AM231153.1 | - | 11 - 12 | 1952 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Col6a4
Genomic Map