Assay Details
| Gene Symbol: Col6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026763.2 | NP_081039.2 | 19 - 20 | 4705 | 64 | |
| GenBank mRNA | AB370265.1 | - | 19 - 20 | 4797 | 64 | |
| AM231153.1 | - | 13 - 14 | 2085 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Col6a4
Genomic Map