| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900024856.mm.1 |
NM_026763.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024856.mm.1 |
NM_026763.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024856.mm.1 |
TC0900003000.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900024859.mm.1 |
NM_026763.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024859.mm.1 |
NM_026763.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024859.mm.1 |
TC0900003000.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900024858.mm.1 |
NM_026763.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024858.mm.1 |
NM_026763.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024858.mm.1 |
TC0900003000.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900024857.mm.1 |
NM_026763.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024857.mm.1 |
NM_026763.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900024857.mm.1 |
TC0900003000.mm.1 |
MTA TC |