Assay Details
| Gene Symbol: Col6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026763.2 | NP_081039.2 | 25 - 26 | 5049 | 72 | |
| GenBank mRNA | AB370265.1 | - | 25 - 26 | 5141 | 72 | |
| AM231153.1 | - | 19 - 20 | 2429 | 72 | ||
More Information
Set Membership: |
|

Col6a4
Genomic Map