Assay Details
| Gene Symbol: Col6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026763.2 | NP_081039.2 | 33 - 34 | 5841 | 73 | |
| GenBank mRNA | AB370265.1 | - | 32 - 33 | 5933 | 73 | |
| AM231153.1 | - | 27 - 28 | 3221 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Col6a4
Genomic Map