Assay Details
| Gene Symbol: Col6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026763.2 | NP_081039.2 | 9 - 10 | 4001 | 79 | |
| GenBank mRNA | AB370265.1 | - | 9 - 10 | 4093 | 79 | |
| AM231153.1 | - | 3 - 4 | 1381 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Col6a4
Genomic Map