Assay Details
| Gene Symbol: Tmem179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028419.1 | 1 - 2 | 136 | 96 | ||
| GenBank mRNA | AK005135.1 | - | 1 - 2 | 125 | 96 | |
| BC061068.1 | - | 1 - 2 | 106 | 96 | ||
| BC087914.1 | - | 1 - 2 | 122 | 96 | ||
| BC144813.1 | - | 1 - 2 | 136 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem179b
Genomic Map