Assay Details
| Gene Symbol: Rfx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027787.1 | NP_082063.1 | 5 - 6 | 638 | 74 | |
| GenBank mRNA | AK044439.1 | - | 5 - 6 | 638 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfx2
Genomic Map