Assay Details
| Gene Symbol: Vav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163815.1 | NP_001157287.1 | 14 - 15 | 1295 | 91 | |
| NM_001163816.1 | NP_001157288.1 | 13 - 14 | 1367 | 91 | ||
| NM_011691.4 | NP_035821.3 | 13 - 14 | 1367 | 91 | ||
| GenBank mRNA | AK088586.1 | - | 13 - 14 | 1353 | 91 | |
| AK146347.1 | - | 13 - 14 | 1345 | 91 | ||
| AK150398.1 | - | 13 - 14 | 1363 | 91 | ||
| AK151183.1 | - | 13 - 14 | 1364 | 91 | ||
| AK151214.1 | - | 14 - 15 | 1296 | 91 | ||
| AK151367.1 | - | 13 - 14 | 1365 | 91 | ||
| AK151373.1 | - | 13 - 14 | 1365 | 91 | ||
| AK151831.1 | - | 13 - 14 | 1363 | 91 | ||
| AK152073.1 | - | 13 - 14 | 1365 | 91 | ||
| AK152425.1 | - | 13 - 14 | 1365 | 91 | ||
| AK152555.1 | - | 13 - 14 | 1353 | 91 | ||
| AK153052.1 | - | 13 - 14 | 1312 | 91 | ||
| AK153265.1 | - | 13 - 14 | 1365 | 91 | ||
| AK154183.1 | - | 13 - 14 | 1352 | 91 | ||
| AK159326.1 | - | 13 - 14 | 1365 | 91 | ||
| BC020487.1 | - | 13 - 14 | 1350 | 3965451 | 91 | |
| CT010321.1 | - | 13 - 14 | 1269 | 91 | ||
| X64361.1 | - | 13 - 14 | 1282 | 91 | ||
More Information
Set Membership: |
|

Vav1
Genomic Map