| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700009933.mm.1 |
NM_001163815.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700009933.mm.1 |
NM_001163815.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700009933.mm.1 |
NM_001163816.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700009933.mm.1 |
NM_001163816.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700009933.mm.1 |
NM_011691.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700009933.mm.1 |
NM_011691.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700009933.mm.1 |
TC1700001033.mm.1 |
MTA TC |