Assay Details
| Gene Symbol: Ip6k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005166.1 | - | 1 - 2 | 72 | 97 | |
| AK050593.1 | - | 1 - 2 | 60 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005166.1 transcripts.
Set Membership: |
|

Ip6k2
Genomic Map