Assay Details
| Gene Symbol: Ip6k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029634.2 | NP_083910.2 | 4 - 5 | 869 | 67 | |
| GenBank mRNA | AK050593.1 | - | 4 - 5 | 725 | 67 | |
| BC039922.1 | - | 4 - 5 | 880 | 2609803 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ip6k2
Genomic Map