Assay Details
| Gene Symbol: Slc26a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134420.4 | NP_599252.2 | 1104 | 58 | ||
| GenBank mRNA | AB099881.1 | - | 265 | 58 | ||
| AF248494.1 | - | 450 | 58 | |||
| AK029238.1 | - | 2030 | 58 | |||
| AK046993.1 | - | 2015 | 58 | |||
| AK053088.1 | - | 406 | 58 | |||
| AK080750.1 | - | 463 | 58 | |||
| AK163670.1 | - | 1105 | 58 | |||
| AK163671.1 | - | 1105 | 58 | |||
| AY032863.1 | - | 409 | 58 | |||
| AY049076.1 | - | 364 | 58 | |||
| BC028856.1 | - | 428 | 4165725 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a6
Genomic Map