Assay Details
| Gene Symbol: Slc26a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134420.4 | NP_599252.2 | 1763 | 71 | ||
| GenBank mRNA | AB099881.1 | - | 924 | 71 | ||
| AF248494.1 | - | 1109 | 71 | |||
| AK029238.1 | - | 2685 | 71 | |||
| AK046993.1 | - | 3023 | 71 | |||
| AK053088.1 | - | 1414 | 71 | |||
| AK080750.1 | - | 1067 | 71 | |||
| AK163670.1 | - | 1764 | 71 | |||
| AK163671.1 | - | 1764 | 71 | |||
| AY032863.1 | - | 1068 | 71 | |||
| AY049076.1 | - | 1023 | 71 | |||
| BC028856.1 | - | 1087 | 4165725 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a6
Genomic Map