Assay Details
| Gene Symbol: Sema3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042779.2 | NP_001036244.1 | 3 - 4 | 318 | 86 | |
| NM_001291537.1 | NP_001278466.1 | 3 - 4 | 582 | 86 | ||
| NM_001291538.1 | NP_001278467.1 | 3 - 4 | 576 | 86 | ||
| NM_001291539.1 | NP_001278468.1 | 2 - 3 | 259 | 86 | ||
| NR_111986.1 | 1 - 2 | 138 | 86 | |||
| GenBank mRNA | AK014333.1 | - | 1 - 2 | 74 | 86 | |
| AK033597.1 | - | 2 - 3 | 269 | 86 | ||
| AK037034.1 | - | 3 - 4 | 307 | 86 | ||
| AK043634.1 | - | 2 - 3 | 212 | 86 | ||
| AK048364.1 | - | 2 - 3 | 540 | 86 | ||
| BC090669.1 | - | 2 - 3 | 239 | 30696359 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC090669.1, NM_001042779.2, NM_001291537.1, NM_001291538.1, NM_001291539.1 transcripts.
Set Membership: |
|

Sema3b
Genomic Map