Assay Details
| Gene Symbol: Hyal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178020.3 | NP_821139.2 | 1 - 2 | 42 | 68 | |
| GenBank mRNA | AY048681.1 | - | 1 - 2 | 31 | 68 | |
| BC018457.1 | - | 1 - 2 | 42 | 3592874 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY048681.1, BC018457.1, NM_178020.3 transcripts.
Set Membership: |
|

Hyal3
Genomic Map