Assay Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286684.1 | NP_001273613.1 | 11 - 12 | 2907 | 140 | |
| GenBank mRNA | AF004666.1 | - | 11 - 12 | 2902 | 140 | |
More Information
Set Membership: |
|

Slc8a1
Genomic Map