Assay Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112798.2 | NP_001106269.1 | 1 - 2 | 82 | 75 | |
| NM_001286684.1 | NP_001273613.1 | 1 - 2 | 82 | 75 | ||
| NM_011406.3 | NP_035536.2 | 1 - 2 | 82 | 75 | ||
| NR_104580.1 | 1 - 2 | 82 | 75 | |||
| GenBank mRNA | AF004666.1 | - | 1 - 2 | 77 | 75 | |
| BB660129.1 | - | 1 - 2 | 82 | 75 | ||
| U70033.1 | - | 1 - 2 | 72 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF004666.1, NM_001112798.2, NM_001286684.1, NM_011406.3, U70033.1 transcripts.
Set Membership: |
|

Slc8a1
Genomic Map