Assay Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112798.2 | NP_001106269.1 | 2 - 3 | 1902 | 75 | |
| NM_001286684.1 | NP_001273613.1 | 2 - 3 | 1902 | 75 | ||
| NM_011406.3 | NP_035536.2 | 2 - 3 | 1902 | 75 | ||
| NR_104580.1 | 2 - 3 | 1902 | 75 | |||
| GenBank mRNA | AK048160.1 | - | 1469 | 75 | ||
| BC079673.1 | - | 1 - 2 | 1793 | 30532156 | 75 | |
| DQ388998.1 | - | 1 - 2 | 1792 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc8a1
Genomic Map