Assay Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286684.1 | NP_001273613.1 | 4 - 5 | 2038 | 70 | |
| NM_011406.3 | NP_035536.2 | 4 - 5 | 2038 | 70 | ||
| GenBank mRNA | AF004666.1 | - | 4 - 5 | 2033 | 70 | |
| U70033.1 | - | 4 - 5 | 2028 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc8a1
Genomic Map