Assay Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112798.2 | NP_001106269.1 | 7 - 8 | 2334 | 65 | |
| NM_001286684.1 | NP_001273613.1 | 9 - 10 | 2370 | 65 | ||
| NM_011406.3 | NP_035536.2 | 9 - 10 | 2370 | 65 | ||
| NR_104580.1 | 7 - 8 | 2369 | 65 | |||
| GenBank mRNA | AK048160.1 | - | 1832 | 65 | ||
| BC079673.1 | - | 6 - 7 | 2225 | 30532156 | 65 | |
| DQ388998.1 | - | 6 - 7 | 2259 | 65 | ||
| U70033.1 | - | 9 - 10 | 2360 | 65 | ||
More Information
Set Membership: |
|

Slc8a1
Genomic Map